siRNA Details
Gene Symbol: MIIP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021933.3 | NP_068752.2 | 6, 7 | 1103 | ||
XM_005263487.3 | XP_005263544.1 | 6, 7 | 932 | |||
XM_011541895.1 | XP_011540197.1 | 6, 7 | 934 | |||
XM_011541896.1 | XP_011540198.1 | 6, 7 | 827 | |||
GenBank mRNA | AK022500.1 | - | 6, 7 | 883 | ||
AK024020.1 | - | 6, 7 | 822 | |||
BC008068.2 | - | 6, 7 | 812 | 3050071 | ||
FJ618905.1 | - | 6, 7 | 1103 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
