siRNA Details
Gene Symbol: FCGR2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136219.1 | NP_001129691.1 | 7 | 1571 | ||
NM_021642.3 | NP_067674.2 | 7 | 1568 | |||
XM_011509291.1 | XP_011507593.1 | 6 | 1518 | |||
XM_017000667.1 | XP_016856156.1 | 6 | 1433 | |||
XM_017000668.1 | XP_016856157.1 | 6 | 1430 | |||
GenBank mRNA | AK225601.1 | - | 7 | 1525 | ||
AK298734.1 | - | 6 | 1131 | |||
J03619.1 | - | 7 | 1523 | |||
M31932.1 | - | 7 | 1525 | |||
X62572.1 | - | 7 | 1505 |
Gene Symbol: FCGR2C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_201563.5 | NP_963857.3 | 7 | 1632 | ||
NR_047648.1 | 7 | 1632 |
Gene Symbol: UFL1-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BM126999.1 | - | 6 | 451 | ||
BM127608.1 | - | 6 | 451 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
