siRNA Details
Gene Symbol: CHIA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001258001.1 | NP_001244930.1 | 5, 6 | 453 | ||
NM_001258003.1 | NP_001244932.1 | 4, 5 | 360 | |||
NM_021797.3 | NP_068569.2 | 3, 4 | 330 | |||
NM_201653.3 | NP_970615.2 | 6, 7 | 655 | |||
XM_017001048.1 | XP_016856537.1 | 4, 5 | 363 | |||
GenBank mRNA | AB025008.1 | - | 3, 4 | 320 | ||
AF290004.1 | - | 6, 7 | 591 | |||
AK290702.1 | - | 5, 6 | 455 | |||
AY789444.1 | - | 5, 6 | 389 | |||
AY825504.1 | - | 4, 5 | 350 | |||
BC036339.1 | - | 3, 4 | 268 | 5184463 | ||
BC047336.1 | - | 4, 5 | 277 | 5185486 | ||
BC106910.1 | - | 2, 3 | 236 | 40031872 | ||
BC139901.1 | - | 4, 5 | 282 | |||
BC139920.1 | - | 4, 5 | 282 |
Gene Symbol: CHIAP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003928.2 | 3, 4 | 1129 | |||
GenBank mRNA | BC031662.2 | - | 3, 4 | 1129 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
