siRNA Details
Gene Symbol: AXL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001699.5 | NP_001690.2 | 2 | 400 | ||
NM_021913.4 | NP_068713.2 | 2 | 400 | |||
GenBank mRNA | AA021223.1 | - | 2 | 403 | ||
AK310881.1 | - | 2 | 221 | |||
AL556721.3 | - | 2 | 396 | |||
BC032229.1 | - | 2 | 371 | 5205825 | ||
DA478638.1 | - | 2 | 257 | |||
HY045208.1 | - | 2 | 375 | |||
M76125.1 | - | 2 | 364 | |||
S65125.1 | - | 2 | 241 | |||
X57019.1 | - | 2 | 241 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
