siRNA Details
Gene Symbol: TRIM34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003827.1 | NP_001003827.1 | 4 | 690 | ||
NM_021616.5 | NP_067629.2 | 4 | 694 | |||
NM_130390.2 | NP_569074.2 | 3 | 610 | |||
GenBank mRNA | AB039902.1 | - | 4 | 698 | ||
AB039904.1 | - | 3 | 592 | |||
AF220143.1 | - | 4 | 677 | |||
AF220144.1 | - | 5 | 610 | |||
AK027876.1 | - | 4 | 666 | |||
AL583914.1 | - | 3 | 299 |
Gene Symbol: TRIM6-TRIM34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003819.3 | NP_001003819.1 | 10 | 1858 | ||
GenBank mRNA | AB039903.1 | - | 10 | 1766 | ||
BC136871.1 | - | 10 | 1777 | 9020862 | ||
BC140722.1 | - | 10 | 1777 | 9021590 | ||
BC144424.1 | - | 10 | 1777 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
