siRNA Details
Gene Symbol: LOC728392 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001162371.2 | NP_001155843.1 | 2 | 1335 | ||
GenBank mRNA | AF217966.1 | - | 2 | 1321 | ||
AI371970.1 | - | 1 | 120 | |||
BC016965.1 | - | 2 | 793 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
