siRNA Details
Gene Symbol: DNAJC12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021800.2 | NP_068572.1 | 4 | 557 | ||
XM_017016431.1 | XP_016871920.1 | 3 | 329 | |||
XM_017016432.1 | XP_016871921.1 | 3 | 300 | |||
GenBank mRNA | AF176012.1 | - | 4 | 542 | ||
AK289537.1 | - | 4 | 507 | |||
BC017018.1 | - | 4 | 493 | 3884093 | ||
CR457184.1 | - | 4 | 389 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
