siRNA Details
Gene Symbol: SLC22A23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286455.1 | NP_001273384.1 | 10 | 5508 | ||
NM_015482.1 | NP_056297.1 | 10 | 5967 | |||
NM_021945.6 | NP_068764.3 | 11 | 5753 | |||
NR_104448.1 | 12 | 5224 | ||||
XM_011514801.2 | XP_011513103.1 | 11 | 6054 | |||
XM_017011180.1 | XP_016866669.1 | 11 | 6531 | |||
XM_017011186.1 | XP_016866675.1 | 11 | 5450 | |||
GenBank mRNA | AJ420525.1 | - | 1 | 1801 | ||
AK025033.1 | - | 1 | 2018 | |||
AL512737.1 | - | 1 | 1639 | |||
BC022217.1 | - | 1 | 3136 | |||
BC090851.1 | - | 1 | 3040 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
