siRNA Details
Gene Symbol: PRKACB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242857.2 | NP_001229786.1 | 13 | 2050 | ||
NM_001242858.2 | NP_001229787.1 | 10 | 1993 | |||
NM_001242859.2 | NP_001229788.1 | 12 | 2041 | |||
NM_001242860.2 | NP_001229789.1 | 13 | 2185 | |||
NM_001242861.2 | NP_001229790.1 | 11 | 2077 | |||
NM_001242862.2 | NP_001229791.1 | 9 | 1789 | |||
NM_002731.3 | NP_002722.1 | 10 | 2151 | |||
NM_182948.3 | NP_891993.1 | 10 | 2055 | |||
XM_005271015.3 | XP_005271072.1 | 12 | 2002 | |||
XM_005271016.1 | XP_005271073.1 | 12 | 2127 | |||
XM_005271017.3 | XP_005271074.1 | 11 | 1992 | |||
XM_005271018.4 | XP_005271075.1 | 11 | 2312 | |||
XM_005271019.1 | XP_005271076.1 | 10 | 2068 | |||
XM_006710758.1 | XP_006710821.1 | 12 | 2141 | |||
GenBank mRNA | AB209189.1 | - | 13 | 2120 | ||
AK091420.1 | - | 11 | 1751 | |||
BC035058.1 | - | 10 | 2069 | 5262372 | ||
BX537705.1 | - | 10 | 2032 | |||
BX641026.1 | - | 10 | 2031 | |||
CR936631.1 | - | 12 | 1844 | |||
M34181.1 | - | 10 | 1845 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
