siRNA Details
Gene Symbol: CXorf56 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170569.1 | NP_001164040.1 | 3 | 419 | ||
NM_022101.3 | NP_071384.1 | 3 | 337 | |||
GenBank mRNA | AK096297.1 | - | 3 | 383 | ||
BC023506.2 | - | 3 | 298 | |||
BQ223940.1 | - | 3 | 302 | |||
DA590933.1 | - | 3 | 421 | |||
DA921725.1 | - | 3 | 312 | |||
DB033119.1 | - | 3 | 352 |
