siRNA Details
Gene Symbol: SERF1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021967.2 | NP_068802.1 | 2 | 292 | ||
NM_022968.1 | NP_075257.1 | 2 | 292 | |||
GenBank mRNA | AF073518.1 | - | 2 | 259 | ||
AF073519.1 | - | 2 | 268 | |||
BC021174.1 | - | 2 | 291 | 4824358 | ||
BC035932.1 | - | 2 | 221 | 4701505 | ||
BC093880.1 | - | 2 | 108 | 7939725 | ||
BC093882.1 | - | 2 | 104 | 7939727 | ||
DB444407.1 | - | 2 | 294 |
Gene Symbol: SERF1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001178087.1 | NP_001171558.1 | 2 | 286 | ||
NM_022978.2 | NP_075267.1 | 2 | 286 | |||
GenBank mRNA | BC101970.1 | - | 2 | 106 | 40000410 | |
BC103687.1 | - | 2 | 106 | 40000408 | ||
BC104738.1 | - | 2 | 106 | 40000411 | ||
BC143255.1 | - | 2 | 104 | 9051757 | ||
BC143260.1 | - | 2 | 108 | 9051762 | ||
BC171749.1 | - | 2 | 108 | 9053250 | ||
CD679521.1 | - | 2 | 166 | |||
DB454907.1 | - | 2 | 286 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
