siRNA Details
Gene Symbol: KDM5B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001314042.1 | NP_001300971.1 | 1 | 1026 | ||
NM_006618.4 | NP_006609.3 | 1 | 1026 | |||
XM_011509088.2 | XP_011507390.1 | 1 | 78 | |||
GenBank mRNA | AJ243706.1 | - | 1 | 1029 | ||
DA436869.1 | - | 1 | 78 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
