siRNA Details
Gene Symbol: CXorf56 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170569.1 | NP_001164040.1 | 7 | 2300 | ||
NM_001170570.1 | NP_001164041.1 | 6 | 2176 | |||
NM_022101.3 | NP_071384.1 | 7 | 2218 | |||
GenBank mRNA | BC023506.2 | - | 8 | 1158 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
