siRNA Details
Gene Symbol: DIO3OS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF469199.1 | - | 3 | 852 | ||
AF469200.1 | - | 4 | 698 | |||
AF469205.1 | - | 1 | 83 | |||
BC065701.1 | - | 3 | 1867 | |||
BC111049.1 | - | 2 | 1684 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
