siRNA Details
Gene Symbol: CRLF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022148.3 | NP_071431.2 | 2 | 255 | ||
NR_110830.1 | 2 | 252 | ||||
XM_011546181.2 | XP_011544483.1 | 2 | 290 | |||
XM_011546182.2 | XP_011544484.1 | 2 | 294 | |||
GenBank mRNA | AB052639.1 | - | 2 | 166 | ||
AF142570.1 | - | 2 | 182 | |||
AF338733.1 | - | 2 | 166 | |||
AK026800.1 | - | 2 | 254 | |||
AK223303.1 | - | 2 | 282 | |||
AK225583.1 | - | 2 | 254 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
