siRNA Details
Gene Symbol: PDE4DIP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002810.3 | NP_001002810.1 | 4 | 910 | ||
NM_001195260.1 | NP_001182189.1 | 5 | 607 | |||
NM_001195261.1 | NP_001182190.1 | 5 | 705 | |||
NM_022359.5 | NP_071754.3 | 7 | 1343 | |||
GenBank mRNA | AB042557.1 | - | 4 | 786 | ||
BC025406.1 | - | 7 | 1354 | |||
BC152553.1 | - | 6 | 3698 | |||
BI550399.1 | - | 5 | 624 |
Gene Symbol: LOC653513 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017003026.1 | XP_016858515.1 | 7 | 1357 | ||
XM_017003033.1 | XP_016858522.1 | 7 | 1192 | |||
XM_017003040.1 | XP_016858529.1 | 5 | 700 | |||
GenBank mRNA | AJ708549.1 | - | 1 | 218 | ||
EL953210.1 | - | 3 | 525 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
