siRNA Details
Gene Symbol: PJA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022368.4 | NP_071763.2 | 2 | 458 | ||
NM_145119.3 | NP_660095.1 | 2 | 458 | |||
XM_011531011.2 | XP_011529313.1 | 2 | 265 | |||
GenBank mRNA | AF262024.1 | - | 1 | 387 | ||
AF264620.1 | - | 2 | 373 | |||
AK096611.1 | - | 2 | 457 | |||
AK290543.1 | - | 2 | 358 | |||
BC075803.1 | - | 2 | 365 | 3913310 | ||
BC105051.1 | - | 1 | 147 | 8144054 | ||
BC105053.1 | - | 1 | 147 | 8144056 | ||
DA778001.1 | - | 2 | 457 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
