siRNA Details
Gene Symbol: RAB3IP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022456.4 | NP_071901.2 | 2 | 541 | ||
NM_175623.3 | NP_783322.1 | 2 | 675 | |||
NM_175624.3 | NP_783323.1 | 2 | 541 | |||
NM_175625.3 | NP_783324.1 | 2 | 675 | |||
NR_103519.1 | 2 | 541 | ||||
NR_103520.1 | 2 | 675 | ||||
XM_006719226.3 | XP_006719289.3 | 2 | 664 | |||
XM_006719227.3 | XP_006719290.3 | 2 | 337 | |||
XM_017018773.1 | XP_016874262.1 | 2 | 299 | |||
XM_017018775.1 | XP_016874264.1 | 2 | 337 | |||
GenBank mRNA | AJ312896.1 | - | 2 | 365 | ||
AJ312897.1 | - | 2 | 657 | |||
AJ312898.1 | - | 2 | 365 | |||
AJ312899.1 | - | 2 | 657 | |||
AJ312900.1 | - | 2 | 365 | |||
AJ312901.1 | - | 2 | 657 | |||
AK026201.1 | - | 2 | 365 | |||
AK057844.1 | - | 2 | 270 | |||
AK225441.1 | - | 2 | 365 | |||
AK314547.1 | - | 2 | 376 | |||
BC059358.1 | - | 2 | 330 | 5300636 | ||
DA211792.1 | - | 2 | 376 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
