siRNA Details
Gene Symbol: CDH24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022478.3 | NP_071923.2 | 14 | 3448 | ||
NM_144985.3 | NP_659422.2 | 13 | 3334 | |||
GenBank mRNA | AK025342.1 | - | 6 | 1752 | ||
AL137477.1 | - | 5 | 1907 | |||
AY358199.1 | - | 13 | 3290 | |||
BC094810.1 | - | 2 | 1212 | |||
BC110867.1 | - | 6 | 1904 | |||
BX110381.1 | - | 2 | 503 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
