siRNA Details
Gene Symbol: CRLF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011546181.2 | XP_011544483.1 | 8 | 1633 | ||
GenBank mRNA | AF142570.1 | - | 8, 9 | 1525 | ||
AK026800.1 | - | 8 | 1779 | |||
AK225583.1 | - | 8 | 1779 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
