siRNA Details
Gene Symbol: HERPUD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022373.4 | NP_071768.3 | 5, 6 | 1160 | ||
XM_006715765.1 | XP_006715828.1 | 3, 4 | 968 | |||
XM_011515473.1 | XP_011513775.1 | 2, 3 | 1166 | |||
XM_017012506.1 | XP_016867995.1 | 4, 5 | 1358 | |||
GenBank mRNA | AK025966.1 | - | 16 | |||
AK055708.1 | - | 4, 5 | 1307 | |||
AK315685.1 | - | 4, 5 | 784 | |||
BC005091.1 | - | 5, 6 | 966 | 3840918 | ||
BC020264.1 | - | 5, 6 | 1117 | 4878523 |
