siRNA Details
Gene Symbol: VPS16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022575.3 | NP_072097.2 | 15, 16 | 1562 | ||
NM_080413.2 | NP_536338.1 | 11, 12 | 1130 | |||
GenBank mRNA | AF308801.1 | - | 15, 16 | 1498 | ||
AK055787.1 | - | 11, 12 | 1100 | |||
AL834401.1 | - | 11, 12 | 1050 | |||
BC012422.1 | - | 3, 4 | 213 | |||
BC021291.2 | - | 1, 2 | 81 | |||
BC073959.1 | - | 3, 4 | 632 | 6182570 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
