siRNA Details
Gene Symbol: EBF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022659.3 | NP_073150.2 | 10 | 1264 | ||
GenBank mRNA | AK021562.1 | - | 3 | 303 | ||
AK304372.1 | - | 9 | 912 | |||
AY700779.1 | - | 2 | 192 | |||
BC069665.1 | - | 3 | 281 | |||
BC069726.1 | - | 3 | 284 | |||
BC069747.1 | - | 3 | 283 | |||
BC069768.1 | - | 3 | 284 | |||
BC074794.2 | - | 2 | 220 | |||
BC113478.1 | - | 2 | 248 | |||
BC113504.1 | - | 2 | 248 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
