siRNA Details
Gene Symbol: PLPPR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022737.2 | NP_073574.2 | 4 | 525 | ||
XM_005260017.1 | XP_005260074.1 | 4 | 525 | |||
XM_017027147.1 | XP_016882636.1 | 4 | 528 | |||
GenBank mRNA | AK023117.1 | - | 4 | 486 | ||
AK225482.1 | - | 4 | 440 | |||
AY304516.1 | - | 4 | 480 | |||
AY339628.1 | - | 3 | 180 | |||
BC009378.1 | - | 4 | 368 | 4125316 |
