siRNA Details
Gene Symbol: S100PBP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256121.1 | NP_001243050.1 | 3, 4 | 1119 | ||
NM_022753.3 | NP_073590.2 | 3, 4 | 1133 | |||
NR_135107.1 | 2, 3 | 961 | ||||
XM_011541961.2 | XP_011540263.1 | 3, 4 | 1068 | |||
XM_011541962.2 | XP_011540264.1 | 3, 4 | 1177 | |||
XM_011541963.1 | XP_011540265.1 | 3, 4 | 1133 | |||
GenBank mRNA | AK022965.1 | - | 1, 2 | 440 | ||
AK090711.1 | - | 3, 4 | 1119 | |||
BC015175.1 | - | 3, 4 | 992 | 3904501 | ||
BX647667.1 | - | 3, 4 | 1060 | |||
BX647916.1 | - | 3, 4 | 1097 | |||
KU178750.1 | - | 1, 2 | 842 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
