siRNA Details
Gene Symbol: DGCR8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190326.1 | NP_001177255.1 | 4 | 1429 | ||
NM_022720.6 | NP_073557.3 | 4 | 1429 | |||
XM_006724268.2 | XP_006724331.1 | 1 | 32 | |||
GenBank mRNA | AK025539.1 | - | 3 | 393 | ||
AK222796.1 | - | 3 | 393 | |||
BC009323.2 | - | 2 | 232 | |||
BC037564.1 | - | 4 | 1379 | |||
BC078147.1 | - | 3 | 1143 | 6014346 | ||
BX649187.1 | - | 2 | 157 | |||
CR456356.1 | - | 3 | 1087 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
