siRNA Details
Gene Symbol: LOC285074 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC069048.1 | - | 2 | 306 | ||
BC132791.1 | - | 2 | 265 | |||
BC132793.1 | - | 2 | 265 | |||
BM454994.1 | - | 2 | 477 | |||
BX353812.2 | - | 2 | 525 |
Gene Symbol: RMND5A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022780.3 | NP_073617.1 | 2 | 548 | ||
GenBank mRNA | AK022815.1 | - | 2 | 525 | ||
BC012165.1 | - | 2 | 269 | 4636136 | ||
BC047668.1 | - | 2 | 478 | 6066798 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
