siRNA Details
Gene Symbol: CLEC7A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022570.4 | NP_072092.2 | 1, 2 | 298 | ||
NM_197947.2 | NP_922938.1 | 1, 2 | 298 | |||
NM_197948.2 | NP_922939.1 | 1, 2 | 298 | |||
NM_197949.2 | NP_922940.1 | 1, 2 | 298 | |||
NM_197954.2 | NP_922945.1 | 1, 2 | 298 | |||
NR_125336.1 | 1, 2 | 298 | ||||
XM_006719135.3 | XP_006719198.1 | 1, 2 | 293 | |||
XM_017019822.1 | XP_016875311.1 | 1, 2 | 292 | |||
XM_017019823.1 | XP_016875312.1 | 1, 2 | 195 | |||
GenBank mRNA | AF313468.1 | - | 2, 3 | 162 | ||
AF313469.1 | - | 1, 2 | 161 | |||
AF400595.1 | - | 1, 2 | 111 | |||
AF400596.1 | - | 1, 2 | 111 | |||
AF400597.1 | - | 1, 2 | 111 | |||
AF400598.1 | - | 1, 2 | 111 | |||
AF400601.1 | - | 1, 2 | 111 | |||
AF400602.1 | - | 1, 2 | 111 | |||
AJ312372.1 | - | 1, 2 | 266 | |||
AJ312373.1 | - | 1, 2 | 266 | |||
AK294111.1 | - | 1, 2 | 290 | |||
AK295108.1 | - | 1, 2 | 242 | |||
AK297028.1 | - | 1, 2 | 182 | |||
AK298679.1 | - | 1, 2 | 214 | |||
AK313247.1 | - | 1, 2 | 229 | |||
AY009090.2 | - | 1, 2 | 182 | |||
AY026769.2 | - | 1, 2 | 111 | |||
AY026770.2 | - | 1, 2 | 111 | |||
AY026771.1 | - | 1, 2 | 111 | |||
AY359002.1 | - | 1, 2 | 270 | |||
BC013385.1 | - | 1, 2 | 167 | |||
BC071746.1 | - | 1, 2 | 212 | 4690475 | ||
BC093829.1 | - | 1, 2 | 187 | |||
BC093831.1 | - | 1, 2 | 187 | |||
CR749536.1 | - | 1, 2 | 241 | |||
DA193873.1 | - | 1, 2 | 298 | |||
KU178745.1 | - | 1, 2 | 111 | |||
KU178746.1 | - | 1, 2 | 111 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
