siRNA Details
Gene Symbol: PLPPR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170635.1 | NP_001164106.1 | 10 | 1804 | ||
NM_022737.2 | NP_073574.2 | 10 | 1863 | |||
XM_005260017.1 | XP_005260074.1 | 10 | 1879 | |||
XM_017027147.1 | XP_016882636.1 | 10 | 1882 | |||
XM_017027148.1 | XP_016882637.1 | 10 | 1800 | |||
GenBank mRNA | AK023117.1 | - | 10 | 1824 | ||
AK094869.1 | - | 10 | 1692 | |||
AK225482.1 | - | 9 | 1301 | |||
AL353953.1 | - | 2 | 502 | |||
AY304516.1 | - | 10 | 1818 | |||
AY339628.1 | - | 9 | 1518 | |||
BC009378.1 | - | 10 | 1706 | 4125316 | ||
CR936652.1 | - | 5 | 2269 |
