siRNA Details
Gene Symbol: FAM129B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001035534.2 | NP_001030611.1 | 4 | 400 | ||
NM_022833.3 | NP_073744.2 | 4 | 612 | |||
XM_005252135.2 | XP_005252192.2 | 5 | 459 | |||
XM_011518925.1 | XP_011517227.1 | 5 | 631 | |||
GenBank mRNA | AB210016.1 | - | 4 | 350 | ||
AF151783.1 | - | 4 | 382 | |||
AF192911.1 | - | 4 | 603 | |||
AL137555.1 | - | 3 | 220 | |||
BM557756.1 | - | 574 | ||||
BX280720.1 | - | 4 | 486 | |||
CN423640.1 | - | 4 | 402 |
