siRNA Details
Gene Symbol: CLEC7A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022570.4 | NP_072092.2 | 5 | 1200 | ||
NM_197947.2 | NP_922938.1 | 6 | 1338 | |||
NM_197948.2 | NP_922939.1 | 5 | 1219 | |||
NM_197949.2 | NP_922940.1 | 4 | 1081 | |||
NM_197950.2 | NP_922941.1 | 4 | 1101 | |||
NR_125336.1 | 7 | 1425 | ||||
XM_017019822.1 | XP_016875311.1 | 6 | 1296 | |||
XM_017019823.1 | XP_016875312.1 | 5 | 1061 | |||
GenBank mRNA | AF313469.1 | - | 5 | 1063 | ||
AY009090.2 | - | 5 | 1084 | |||
AY359002.1 | - | 5 | 1172 | |||
CR749536.1 | - | 6 | 2407 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
