siRNA Details
Gene Symbol: MARC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022746.3 | NP_073583.3 | 7 | 1697 | ||
XM_011509900.2 | XP_011508202.1 | 7 | 1748 | |||
XM_011509903.2 | XP_011508205.1 | 7 | 2124 | |||
XM_011509904.2 | XP_011508206.1 | 7 | 1401 | |||
XM_017002096.1 | XP_016857585.1 | 7 | 1277 | |||
XM_017002097.1 | XP_016857586.1 | 7 | 1330 | |||
GenBank mRNA | AK000035.1 | - | 4 | 719 | ||
AK026043.1 | - | 7 | 1493 | |||
AK092439.1 | - | 6 | 2168 | |||
AK094105.1 | - | 8 | 1411 | |||
AK225416.1 | - | 7 | 1493 | |||
AK290812.1 | - | 7 | 1469 | |||
BC010619.1 | - | 7 | 1465 | 3872779 | ||
CB048738.1 | - | 1 | 459 |
