siRNA Details
Gene Symbol: S100PBP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256121.1 | NP_001243050.1 | 7 | 3762 | ||
NM_022753.3 | NP_073590.2 | 7 | 3776 | |||
NR_135106.1 | 8 | 3675 | ||||
NR_135107.1 | 7 | 3632 | ||||
XM_011541961.2 | XP_011540263.1 | 7 | 3711 | |||
XM_011541962.2 | XP_011540264.1 | 7 | 3820 | |||
XM_017002098.1 | XP_016857587.1 | 7 | 3773 | |||
GenBank mRNA | AK022965.1 | - | 6 | 3110 | ||
AK025811.1 | - | 1 | 1462 | |||
BX647916.1 | - | 7 | 3739 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
