siRNA Details
Gene Symbol: SFXN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001322977.1 | NP_001309906.1 | 11 | 4030 | ||
NM_001322978.1 | NP_001309907.1 | 11 | 3973 | |||
NM_001322980.1 | NP_001309909.1 | 10 | 3874 | |||
NM_001322981.1 | NP_001309910.1 | 10 | 3902 | |||
NM_001322982.1 | NP_001309911.1 | 11 | 4001 | |||
NM_022754.6 | NP_073591.2 | 11 | 4002 | |||
GenBank mRNA | AF327346.1 | - | 12 | 1678 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
