siRNA Details
Gene Symbol: MEAF6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270875.1 | NP_001257804.1 | 5, 6 | 614 | ||
GenBank mRNA | AK021792.1 | - | 5, 6 | 549 | ||
AK225434.1 | - | 5, 6 | 549 | |||
AK225598.1 | - | 5, 6 | 549 | |||
AY007166.1 | - | 5, 6 | 539 | |||
AY211926.1 | - | 5, 6 | 538 | |||
BC016328.1 | - | 5, 6 | 573 | 4102633 | ||
BC056406.1 | - | 5, 6 | 570 | 6471256 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001290701, BC108404, AK012867.
Gene Ontology Categories:
