siRNA Details
Gene Symbol: MTHFSD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159377.1 | NP_001152849.1 | 8 | 2247 | ||
NM_001159378.1 | NP_001152850.1 | 8 | 2247 | |||
NM_001159379.1 | NP_001152851.1 | 8 | 2244 | |||
NM_001159380.1 | NP_001152852.1 | 8 | 2211 | |||
NM_022764.2 | NP_073601.2 | 8 | 2244 | |||
NR_027489.1 | 9 | 2317 | ||||
NR_027490.1 | 8 | 2307 | ||||
XM_005256101.1 | XP_005256158.1 | 8 | 2287 | |||
XM_005256105.1 | XP_005256162.1 | 7 | 2202 | |||
XM_005256106.1 | XP_005256163.1 | 7 | 2111 | |||
XM_006721247.3 | XP_006721310.1 | 7 | 2118 | |||
XM_011523280.2 | XP_011521582.1 | 8 | 2287 | |||
XM_011523285.1 | XP_011521587.1 | 9 | 2285 | |||
XM_011523286.1 | XP_011521588.1 | 9 | 2385 | |||
XM_011523287.1 | XP_011521589.1 | 9 | 2289 | |||
XM_011523288.1 | XP_011521590.1 | 8 | 2217 | |||
XM_011523289.1 | XP_011521591.1 | 8 | 2294 | |||
XM_017023571.1 | XP_016879060.1 | 7 | 2133 | |||
XM_017023572.1 | XP_016879061.1 | 7 | 2130 | |||
XM_017023573.1 | XP_016879062.1 | 7 | 2110 | |||
XM_017023574.1 | XP_016879063.1 | 8 | 2210 | |||
GenBank mRNA | AK023060.1 | - | 8 | 2222 | ||
AK096836.1 | - | 9 | 2318 | |||
AK125015.1 | - | 1 | 2836 |
