siRNA Details
Gene Symbol: DGCR8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190326.1 | NP_001177255.1 | 13 | 3190 | ||
NM_022720.6 | NP_073557.3 | 14 | 3289 | |||
XM_006724268.2 | XP_006724331.1 | 11 | 1924 | |||
GenBank mRNA | AK025427.1 | - | 2 | 728 | ||
AK025539.1 | - | 13 | 2253 | |||
AK025780.1 | - | 9 | 1565 | |||
AK094381.1 | - | 2 | 2079 | |||
AY189282.1 | - | 10 | 1583 | |||
BC009323.2 | - | 12 | 2092 | |||
BC037564.1 | - | 14 | 3239 | |||
BC078147.1 | - | 12 | 2904 | 6014346 | ||
BX649187.1 | - | 11 | 2181 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
