siRNA Details
Gene Symbol: RFX7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022841.5 | NP_073752.5 | 9 | 4338 | ||
XM_005254603.3 | XP_005254660.2 | 10 | 4432 | |||
XM_011521925.2 | XP_011520227.1 | 9 | 4270 | |||
XM_017022506.1 | XP_016877995.1 | 10 | 7155 | |||
XM_017022507.1 | XP_016877996.1 | 9 | 4398 | |||
XM_017022508.1 | XP_016877997.1 | 8 | 5567 | |||
GenBank mRNA | AK023056.1 | - | 1 | 2633 | ||
AK024757.1 | - | 1 | 592 | |||
AK126103.1 | - | 2 | 3490 | |||
AL832584.1 | - | 1 | 3148 | |||
AL833367.1 | - | 1 | 2565 | |||
AL834302.1 | - | 1 | 1147 | |||
BC112936.1 | - | 7 | 4158 | 6014850 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001033536, AK037109, AK162159.
Gene Ontology Categories:
