siRNA Details
Gene Symbol: CDCP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022842.4 | NP_073753.3 | 9 | 4741 | ||
XM_011534024.1 | XP_011532326.1 | 9 | 4699 | |||
XM_017007070.1 | XP_016862559.1 | 8 | 4639 | |||
GenBank mRNA | AF468010.1 | - | 9 | 4716 | ||
AK023834.1 | - | 7 | 4318 | |||
AY026461.1 | - | 9 | 4687 | |||
AY167484.1 | - | 9 | 4698 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
