siRNA Details
Gene Symbol: STAG3L4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040585.1 | 6 | 851 | |||
NR_040586.1 | 5 | 775 | ||||
GenBank mRNA | AK023257.1 | - | 5 | 769 | ||
AK307290.1 | - | 6 | 831 | |||
AK308931.1 | - | 6 | 633 | |||
BC026058.2 | - | 5 | 741 | 3936696 |
