siRNA Details
Gene Symbol: VKORC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001311311.1 | NP_001298240.1 | 2 | 494 | ||
NM_024006.5 | NP_076869.1 | 2 | 494 | |||
GenBank mRNA | AF176924.1 | - | 2 | 436 | ||
AK312005.1 | - | 2 | 407 | |||
AY423044.1 | - | 2 | 224 | |||
AY466113.1 | - | 2 | 436 | |||
AY521634.1 | - | 2 | 224 | |||
BC000828.1 | - | 2 | 224 | |||
BC002911.2 | - | 2 | 262 | 3944530 | ||
BI822140.1 | - | 2 | 450 | |||
CN484751.1 | - | 2 | 221 | |||
KU178762.1 | - | 2 | 224 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
