siRNA Details
Gene Symbol: CASD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022900.4 | NP_075051.4 | 10 | 1541 | ||
XM_006716092.3 | XP_006716155.1 | 8 | 1036 | |||
XM_006716093.3 | XP_006716156.2 | 10 | 1379 | |||
XM_011516495.2 | XP_011514797.1 | 10 | 1379 | |||
XM_017012546.1 | XP_016868035.1 | 9 | 1164 | |||
XM_017012547.1 | XP_016868036.1 | 10 | 1322 | |||
GenBank mRNA | AB073397.1 | - | 2 | 314 | ||
AF355594.1 | - | 10 | 1334 | |||
AF397424.2 | - | 10 | 1461 | |||
AK025532.1 | - | 3 | 632 | |||
AK123866.1 | - | 10 | 1379 | |||
AK314329.1 | - | 10 | 1339 | |||
BC063284.1 | - | 10 | 1526 | 5286382 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
