siRNA Details
Gene Symbol: REEP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164730.1 | NP_001158202.1 | 2, 3 | 126 | ||
NM_001164732.1 | NP_001158204.1 | 2, 3 | 242 | |||
NM_022912.2 | NP_075063.1 | 2, 3 | 242 | |||
XM_005264502.1 | XP_005264559.1 | 2, 3 | 242 | |||
XM_005264504.1 | XP_005264561.1 | 2, 3 | 160 | |||
XM_011533044.1 | XP_011531346.1 | 2, 3 | 104 | |||
XM_011533045.2 | XP_011531347.1 | 2, 3 | 89 | |||
XM_017004725.1 | XP_016860214.1 | 2, 3 | 410 | |||
XM_017004726.1 | XP_016860215.1 | 2, 3 | 410 | |||
XM_017004727.1 | XP_016860216.1 | 2, 3 | 410 | |||
GenBank mRNA | AK023172.1 | - | 2, 3 | 242 | ||
AK294300.1 | - | 2, 3 | 223 | |||
AK297201.1 | - | 2, 3 | 177 | |||
AK297287.1 | - | 2, 3 | 126 | |||
AK297359.1 | - | 2, 3 | 214 | |||
AY562239.1 | - | 2, 3 | 98 | |||
BC064846.1 | - | 2, 3 | 204 | 30386342 | ||
CR457301.1 | - | 2, 3 | 98 | |||
DA495719.1 | - | 2, 3 | 126 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
