siRNA Details
Gene Symbol: MTRR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002454.2 | NP_002445.2 | 2 | 153 | ||
NM_024010.2 | NP_076915.2 | 2 | 127 | |||
NR_134480.1 | 2 | 153 | ||||
NR_134481.1 | 2 | 153 | ||||
NR_134482.1 | 2 | 153 | ||||
GenBank mRNA | AF025794.1 | - | 2 | 95 | ||
AF121214.1 | - | 2 | 127 | |||
AK308442.1 | - | 2 | 83 | |||
AK311663.1 | - | 2 | 83 | |||
BC054816.1 | - | 2 | 68 | 5205285 | ||
BC109216.1 | - | 2 | 127 | 40007734 | ||
DA384356.1 | - | 2 | 67 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
