siRNA Details
Gene Symbol: AHNAK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024060.3 | NP_076965.2 | 6 | 791 | ||
GenBank mRNA | BC000926.1 | - | 6 | 733 | ||
BC012477.1 | - | 6 | 727 | |||
CN482151.1 | - | 3 | 171 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
