siRNA Details
Gene Symbol: VANGL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172411.1 | NP_001165882.1 | 8 | 3732 | ||
NM_001172412.1 | NP_001165883.1 | 8 | 3682 | |||
NM_138959.2 | NP_620409.1 | 8 | 3738 | |||
GenBank mRNA | BC000907.2 | - | 1 | 968 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
