siRNA Details
Gene Symbol: C7orf26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024067.3 | NP_076972.2 | 2 | 481 | ||
XM_005249845.4 | XP_005249902.1 | 2 | 499 | |||
XM_011515518.2 | XP_011513820.1 | 2 | 499 | |||
XM_017012615.1 | XP_016868104.1 | 2 | 189 | |||
GenBank mRNA | AK056149.1 | - | 2 | 218 | ||
BI599533.1 | - | 2 | 487 |
