siRNA Details
Gene Symbol: DCAF10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005251577.3 | XP_005251634.1 | 4, 5 | 1164 | ||
XM_017015128.1 | XP_016870617.1 | 3, 4 | 966 | |||
GenBank mRNA | BC003520.1 | - | 1, 2 | 172 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
