siRNA Details
Gene Symbol: PRDM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198.3 | NP_001189.2 | 2 | 482 | ||
XM_006715550.3 | XP_006715613.1 | 2 | 1460 | |||
XM_011536062.2 | XP_011534364.1 | 2 | 1456 | |||
XM_011536063.2 | XP_011534365.1 | 2 | 534 | |||
XM_017011187.1 | XP_016866676.1 | 2 | 269 | |||
GenBank mRNA | AF084199.1 | - | 2 | 363 | ||
AK289556.1 | - | 2 | 426 | |||
AL832963.1 | - | 2 | 416 | |||
AY198414.1 | - | 2 | 482 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
