siRNA Details
Gene Symbol: FAM134A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321109.1 | NP_001308038.1 | 9 | 2516 | ||
NM_001321110.1 | NP_001308039.1 | 9 | 2268 | |||
NM_024293.5 | NP_077269.3 | 9 | 2604 | |||
XM_005246848.2 | XP_005246905.1 | 10 | 2472 | |||
GenBank mRNA | AK091011.1 | - | 10 | 2095 | ||
AL136758.2 | - | 1 | 1151 | |||
BC141851.1 | - | 9 | 2479 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
